Learn More
Cayman Chemical ACTH11-24humnmouser 10mg
SDP
Catalog No. 502943088
Encompass_Preferred
A peptide fragment of ACTH and an MC2R antagonist (IC50 = ~1 nM in HeLa cells expressing the mouse receptor); induces corticosterone and aldosterone production in isolated rat zona glomerulosa and zona fasciculata cells, respectively, with minimum effective concentrations of 4 and 3 µM, respectively; inhibits ACTH-induced stretching, yawning, and penile erections in rats at 1.69 UG/animal, i.c.v.
Catalog No. 50-294-3088 Supplier Cayman Chemical Supplier No. 4155910 MG
A peptide fragment of ACTH and an MC2R antagonist (IC50 = ~1 nM in HeLa cells expressing the mouse receptor); induces corticosterone and aldosterone production in isolated rat zona glomerulosa and zona fasciculata cells, respectively, with minimum effective concentrations of 4 and 3 µM, respectively; inhibits ACTH-induced stretching, yawning, and penile erections in rats at 1.69 UG/animal, i.c.v.