Learn More
Cayman Chemical ACTH4-11humnmousera 10mg
SDP
Catalog No. 502937969
Encompass_Preferred
A peptide fragment of ACTH; induces differentiation of melanocytes at 1,000 nM; inhibits ACE activity in homogenized canine lung (IC50 = 24 µM); has been used to determine substrate specificity of recombinant CPBHz
Catalog No. 50-293-7969 Supplier Cayman Chemical Supplier No. 2743010 MG
A peptide fragment of ACTH; induces differentiation of melanocytes at 1,000 nM; inhibits ACE activity in homogenized canine lung (IC50 = 24 µM); has been used to determine substrate specificity of recombinant CPBHz