Learn More
Cayman Chemical ACTH (7-38) (human) (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502945604
Encompass_Preferred
A peptide fragment of ACTH and an antagonist of MC2R; inhibits ACTH-induced corticosterone production in isolated rat adrenal cells at 0.455 or 4.55 µM; inhibits ACE1 (IC50 = 750 nM for the dog enzyme); inhibits basal cAMP secretion by isolated rat inner adrenocortical cells at 1 µM; induces hypotension in normotensive dogs (ED50 = 0.109 nmol/kg)
Catalog No. 50-294-5604 Supplier Cayman Chemical Supplier No. 4154710 MG
A peptide fragment of ACTH and an antagonist of MC2R; inhibits ACTH-induced corticosterone production in isolated rat adrenal cells at 0.455 or 4.55 µM; inhibits ACE1 (IC50 = 750 nM for the dog enzyme); inhibits basal cAMP secretion by isolated rat inner adrenocortical cells at 1 µM; induces hypotension in normotensive dogs (ED50 = 0.109 nmol/kg)