Learn More
Cytoskeleton, Inc. Acti-Stain 670 Phalloidin, 300 Slides
Catalog No. 50646255
Encompass
Actin staining is very useful in determining the structure and function of the cytoskeleton in living and fixed cells. The actin cytoskeleton is a very dynamic and labile structure in the living cell, but it can be fixed with paraformaldehyde prior to probing or staining for actin structures.
Catalog No. 50-646-255 Supplier Cytoskeleton, Inc. Supplier No. PHDN1A
Actin staining is very useful in determining the structure and function of the cytoskeleton in living and fixed cells. The actin cytoskeleton is a very dynamic and labile structure in the living cell, but it can be fixed with paraformaldehyde prior to probing or staining for actin structures.