Learn More
Biogenex Laboratories Actin : 1A4 : 6mL
SDP
Supplier:  Biogenex Laboratories AM1285M
Encompass_Preferred
Actin is one of two major cytoskeletal proteins. The antibody can be used to identify smooth muscle tumors. It stains leiomyomas and leiomyosarcomas but does not stain carcinomas, melanomas, lymphomas or non-smooth muscle sarcomas. It stains the muscularis and muscularismucosa of the gastrointestinal tract, the uterine myometrium, medial layerof blood vessels, the mesenchymal components of the prostate, andmyoepithelial cells of salivary glands and other organs. The antibody doesnot stain striated muscle such as skeletal and cardiac muscle, endothelium,connective tissue, epithelium or
Catalog No. 50-197-8750