Learn More
Enzo Life Sciences Actin clonal antibody (S12-I) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX810001R100
Encompass_Preferred
Clone: S12-I. Isotype: IgG. Immunogen: Synthetic peptide corresponding to a C-terminal sequence of human β-actin. Species reactivity: Human, Mouse, Rat. Specificity: Recognizes α-actin and β-actin. Applications: ELISA, IP, WB. Purity: Major clone of IgGs obtained from immunoaffinity purified immunoglobulins corresponding to immunogenic peptide. Formulation: Liquid. In 20mM TRIS/HCl, pH 8.0, containing 10mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-0069