Learn More
Bioss Actin Fruit Fly Loading Cont
SDP
Catalog No. 5000065907
Encompass_Preferred
Actin (Fruit Fly Loading Control) Primary Antibody Unconjugated Host Species Rabbit Target Species Clonality Monoclonal Applications WB Concentration 1mg/ml Size 100 ul Storage Buffer 0 01M TBS(pH7 4) with 1% BSA 0 02% Proclin300 and 50% Glycerol
Catalog No. 50-000-65907 Supplier Bioss Supplier No. BSM8777M
Actin (Fruit Fly Loading Control) Primary Antibody Unconjugated Host Species Rabbit Target Species Clonality Monoclonal Applications WB Concentration 1mg/ml Size 100 ul Storage Buffer 0 01M TBS(pH7 4) with 1% BSA 0 02% Proclin300 and 50% Glycerol