Learn More
Biogenex Laboratories Actin, Muscle: HHF35 : 6mL
SDP
Supplier:  Biogenex Laboratories AM0905M
Encompass_Preferred
Actin, a major component of the cytoskeleton, is a globular protein about5 nm in diameter and is composed of one polypeptide chain with a mass ofapproximately 47 kD. This antibody recognizes alpha actin of skeletal,cardiac and smooth muscle cells and gamma actin from smooth musclecells and will stain the cytoplasm in these cells. It is non-reactive withother mesenchymal cells and all epithelial cells except for myoepithelium.It can thus be used as a marker for leiomyomas, leiomyosarcomas,rhabdomyomas and rhabdomyosarcomas..
Catalog No. 50-197-8817