Learn More
Enzo Life Sciences Actin polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADICSA400F
Encompass_Preferred
Host: Rabbit. Immunogen: Bovine skeletal muscle Actin. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Catalog No. 50-200-7184