Learn More
ECM Bioscience Actin Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience AK6510
Encompass_Preferred
The actin phospho-regulation kit can be used to examine phosphorylation of Ty-53 compared to the total expression of actin. The kit includes rabbit polyconal and mouse monoclonal antibodies to detect total actin, and secondary reagents for rabbit polyclonal and mouse monoclonal antibody detection. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms, Ck
Catalog No. 50-206-8602