Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Actin Related Protein 2 Antibody (YA2725) | 10 UL
SDP

Catalog No. 5000443552
Encompass_Preferred

The Actin Related Protein 2 Antibody (YA2725) is a rabbit-derived antibody designed for research applications. It shows reactivity with human, mouse, and rat samples and is suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) techniques.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Clonality: YA2725
  • Molecular weight: 45 kDa
  • Concentration: 0.3 mg/mL
  • Applications: Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF)
  • Storage: Store at -20°C for 1 year. Avoid repeated freeze / thaw cycles.

Catalog No. 50-004-43552 Supplier Medchemexpress LLC Supplier No. HYP8298010UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Actin Related Protein 2 Antibody (YA2725) is a rabbit-derived antibody designed for research applications. It shows reactivity with human, mouse, and rat samples and is suitable for Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF) techniques.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Clonality: YA2725
  • Molecular weight: 45 kDa
  • Concentration: 0.3 mg/mL
  • Applications: Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF)
  • Storage: Store at -20°C for 1 year. Avoid repeated freeze / thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.