Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Actin Related Protein 2 Antibody (YA2725) | 100 UL
SDP

Catalog No. 5000444214
Encompass_Preferred

This Actin Related Protein 2 Antibody (YA2725) is a rabbit-host antibody designed for various research applications. It reacts with human, mouse, and rat samples, making it suitable for a broad range of studies.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Applications: WB (1:500-1:1000), ICC/IF (1:50-1:200)
  • Molecular weight: Predicted and observed at 45 kDa
  • Concentration: 0.3 mg/mL
  • Formulation: Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Storage & Stability: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: With blue ice

Catalog No. 50-004-44214 Supplier Medchemexpress LLC Supplier No. HYP82980100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Actin Related Protein 2 Antibody (YA2725) is a rabbit-host antibody designed for various research applications. It reacts with human, mouse, and rat samples, making it suitable for a broad range of studies.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Applications: WB (1:500-1:1000), ICC/IF (1:50-1:200)
  • Molecular weight: Predicted and observed at 45 kDa
  • Concentration: 0.3 mg/mL
  • Formulation: Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Storage & Stability: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: With blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.