Learn More
Cayman Chemical -ActIn-Term RabbIt Monc 100uL
SDP
Catalog No. 502948154
Encompass_Preferred
An a-AR antagonist and vasoactive compound binds to rat a1A- and a1B-ARs (Kis 4.06 and 6.84 UM respectively) and human platelet a2-ARs (IC50 1 UM) inhibits phenylephrine- clonidine- sypathetic nerve stimulation- or norepinephrine-induced contration of isolated canine saphenous veins from 0.06-60 UM inhibits ADP- collagen- or epinephrine-induced aggregation of isolated human platelets at 100 UM reduces hippocampal neuronal cell death in a rat model of carotid clamping-induced ischemia-reperfusion injury at 10 mg/kg
Catalog No. 50-294-8154 Supplier Cayman Chemical Supplier No. 32127100 UL
An a-AR antagonist and vasoactive compound binds to rat a1A- and a1B-ARs (Kis 4.06 and 6.84 UM respectively) and human platelet a2-ARs (IC50 1 UM) inhibits phenylephrine- clonidine- sypathetic nerve stimulation- or norepinephrine-induced contration of isolated canine saphenous veins from 0.06-60 UM inhibits ADP- collagen- or epinephrine-induced aggregation of isolated human platelets at 100 UM reduces hippocampal neuronal cell death in a rat model of carotid clamping-induced ischemia-reperfusion injury at 10 mg/kg