Learn More
Cayman Chemical -ActIn-Term RabbIt Monc 50uL
SDP
Catalog No. 502946693
Encompass_Preferred
An IKKB NBD mimetic inhibits the protein-protein interaction between NEMO/IKKY and IKKB inhibits TNF-a-induced NF-KB activation in a reporter assay from 25-150 UM reduces LPS-induced IL-6 production in RAW 264.7 cells improves grip strength in the mdx mouse model of Duchenne muscular dystrophy at 30 mg/kg
Catalog No. 50-294-6693 Supplier Cayman Chemical Supplier No. 3237750 UL
An IKKB NBD mimetic inhibits the protein-protein interaction between NEMO/IKKY and IKKB inhibits TNF-a-induced NF-KB activation in a reporter assay from 25-150 UM reduces LPS-induced IL-6 production in RAW 264.7 cells improves grip strength in the mdx mouse model of Duchenne muscular dystrophy at 30 mg/kg