Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Actinine-d9 chloride | 85806-17-3 | 98.0% | 190.72 g/mol | C7H7D9ClNO2 | 5 MG
SDP

Catalog No. 5000264794
Encompass_Preferred

Actinine-d9 chloride is a deuterium-labeled chloride salt of actinine supplied as an isotope-labeled reference standard for analytical research. It is used as an internal standard or tracer in mass spectrometry and metabolite studies for accurate quantitation.

  • Provided as a deuterium-labeled internal standard.
  • Suitable for mass spectrometry quantitation and tracer studies.
  • Molecular formula C7H7D9ClNO2 and molecular weight 190.72 g/mol.
  • Purity (NMR) 98.0% for reliable analytical performance.
  • Recommended storage: 4°C; in solution -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-64794 Supplier Medchemexpress LLC Supplier No. HY113270AS5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Actinine-d9 chloride is a deuterium-labeled chloride salt of actinine supplied as an isotope-labeled reference standard for analytical research. It is used as an internal standard or tracer in mass spectrometry and metabolite studies for accurate quantitation.

  • Provided as a deuterium-labeled internal standard.
  • Suitable for mass spectrometry quantitation and tracer studies.
  • Molecular formula C7H7D9ClNO2 and molecular weight 190.72 g/mol.
  • Purity (NMR) 98.0% for reliable analytical performance.
  • Recommended storage: 4°C; in solution -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.