Learn More
Medchemexpress LLC Actinine-d9 chloride | 85806-17-3 | 98.01% | 190.72 | 10 MG
SDP
Catalog No. 5000437833
Encompass_Preferred
Actinine-d9 chloride is a deuterium-labeled form of Actinine chloride. Actinine chloride is an angiopathic substance produced by gut microbiota from dietary carnitine. This compound is suitable for use as a tracer and as an internal standard in quantitative analyses.
  • Deuterium labeled compound
  • Suitable for use as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS analysis methods
Catalog No. 50-004-37833 Supplier Medchemexpress LLC Supplier No. HY113270AS10MG
Actinine-d9 chloride is a deuterium-labeled form of Actinine chloride. Actinine chloride is an angiopathic substance produced by gut microbiota from dietary carnitine. This compound is suitable for use as a tracer and as an internal standard in quantitative analyses.
  • Deuterium labeled compound
  • Suitable for use as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS analysis methods