Learn More
ATCC Segatella oris
Catalog No. 502381075
Encompass
Segatella oris strain VPI D1A-1A is a whole-genome sequenced bacterial type strain that was isolated from the human oral cavity.
Catalog No. 50-238-1075 Supplier ATCC Supplier No. 33573
Segatella oris strain VPI D1A-1A is a whole-genome sequenced bacterial type strain that was isolated from the human oral cavity.