Learn More
Research Products International Corp Actinomycin D [Actinomycin C1] [Dactinomycin], 10 Milligrams
SDP
Catalog No. 501977636
Encompass_Preferred
Actinomycin D, also known as Dactinomycin, is an antibiotic. 
It works by binding to DNA, inhibiting both DNA and RNA synthesis. This interference with nucleic acid synthesis makes it effective against rapidly dividing cells, such as cancer cells.
Actinomycin D is used as a tool to study DNA and RNA synthesis, cell cycle progression, and cellular responses to DNA damage. Its ability to interfere with nucleic acid synthesis makes it valuable in understanding the molecular mechanisms of cell biology and cancer.
WARNING! This product can expose you to Actinomycin D, a chemical which is known to the State of California to cause cancer and developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7636 Supplier Research Products International Corp Supplier No. A100250.01
Actinomycin D, also known as Dactinomycin, is an antibiotic. 
It works by binding to DNA, inhibiting both DNA and RNA synthesis. This interference with nucleic acid synthesis makes it effective against rapidly dividing cells, such as cancer cells.
Actinomycin D is used as a tool to study DNA and RNA synthesis, cell cycle progression, and cellular responses to DNA damage. Its ability to interfere with nucleic acid synthesis makes it valuable in understanding the molecular mechanisms of cell biology and cancer.
WARNING! This product can expose you to Actinomycin D, a chemical which is known to the State of California to cause cancer and developmental harm. For more information go to www.P65Warnings.ca.gov.