Learn More
Cayman Chemical ActInomycIn D 10mg
SDP
Catalog No. 502954983
Encompass_Preferred
A cyclic polypeptide-containing anticancer antibiotic that binds to DNA and blocks transcription by interfering with RNA synthesis; ~10 nM promotes cell death in a variety of tumors
Catalog No. 50-295-4983 Supplier Cayman Chemical Supplier No. 1142110 MG
A cyclic polypeptide-containing anticancer antibiotic that binds to DNA and blocks transcription by interfering with RNA synthesis; ~10 nM promotes cell death in a variety of tumors