Learn More
Apexbio Technology LLC Actinomycin D, 10mM (in 1mL DMSO). Cas: 50-76-0 MFCD: MFCD00005033. RNA polymerase inhibitor
SDP
Catalog No. 501904941
Encompass_Preferred
IC50: Actinomycin D showed a concentration-dependent decrease of DNA repair activity with the IC50 of 0.42 M [1].Actinomycin D (dactinomycin), a member of actinomycines, which are a class of polypeptide antibiotics isolated from soil bacteria of the genus Streptomyces. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4941 Supplier Apexbio Technology LLC Supplier No. A44485.1
IC50: Actinomycin D showed a concentration-dependent decrease of DNA repair activity with the IC50 of 0.42 M [1].Actinomycin D (dactinomycin), a member of actinomycines, which are a class of polypeptide antibiotics isolated from soil bacteria of the genus Streptomyces. Other sizes are available. Please inqury us for quote.