Learn More
AdipoGen Actinomycin D, 50-76-0, 20 mg
SDP
Catalog No. 501490228
Encompass_Preferred
Actinomycin D (20 mg), AdipoGen Life Sciences. Chemical. CAS: 50-76-0. Formula: C62H86N12O16. MW: 1255.42. Synthetic. Biochemical/physiological actions: An antineoplastic antibiotic that inhibits cell proliferation by forming a stable complex with DNA and blocking the movement of RNA polymerase which interferes with DNA-dependent RNA synthesis. Potent antitumor agent. For cell culture applications, actinomycin D is used as a selection agent and is used in banding techniques to differentiate between different regions of chromosomes.
Catalog No. 50-149-0228 Supplier AdipoGen Supplier No. CDXA0043M020
Actinomycin D (20 mg), AdipoGen Life Sciences. Chemical. CAS: 50-76-0. Formula: C62H86N12O16. MW: 1255.42. Synthetic. Biochemical/physiological actions: An antineoplastic antibiotic that inhibits cell proliferation by forming a stable complex with DNA and blocking the movement of RNA polymerase which interferes with DNA-dependent RNA synthesis. Potent antitumor agent. For cell culture applications, actinomycin D is used as a selection agent and is used in banding techniques to differentiate between different regions of chromosomes.