Learn More
Cayman Chemical Actinomycin D, Purity: ≥95%, CAS: 50-76-0, 25 mg
SDP
Catalog No. 502953639
Encompass_Preferred
A cyclic polypeptide-containing anticancer antibiotic that binds to DNA and blocks transcription by interfering with RNA synthesis; ~10 nM promotes cell death in a variety of tumors
Catalog No. 50-295-3639 Supplier Cayman Chemical Supplier No. 1142125 MG
A cyclic polypeptide-containing anticancer antibiotic that binds to DNA and blocks transcription by interfering with RNA synthesis; ~10 nM promotes cell death in a variety of tumors