Learn More
Gibco™ Actinomycin D
Description
Actinomycin D exhibits high GC selectivity and causes distortion at its binding site. Binding of the nonfluorescent actinomycin D to nucleic acids changes the absorbance of the dye. Actinomycin D has been used for chromosome banding studies.
Cell Analysis, Cell Proliferation, Cell Viability, Proliferation & Function
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Violet |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Colorimetric |
| Form | Solid |
| Product Type | Antibiotic |
| Quantity | 10 mg |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
We recommend reconstituting Actinomycin D in either DMF or DMSO; it should be soluble to at least 1 mM in either solvent.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.