Learn More
Bioworld Actinomycin D, 5 mg
Catalog No. 50255296
Encompass_Preferred
Actinomycin DAntineoplastic antibiotic produced by growth of Streptomyces chrysomallys and other species of Streptomyces.A potent antibiotic that binds to the GpC steps of DNA Technical Information:For in vitro lab use only. Not for human use or consumption.Appearance: SolidBoiling Point : 1386.05 °C at 760 mmHg (Predicted)Density : 1.43 g/cm3 (Predicted)Melting Point: 251-253 °CRTECS Number: AU1575000
Catalog No. 50-255-296 Supplier Bioworld Supplier No. 401100041
Actinomycin DAntineoplastic antibiotic produced by growth of Streptomyces chrysomallys and other species of Streptomyces.A potent antibiotic that binds to the GpC steps of DNA Technical Information:For in vitro lab use only. Not for human use or consumption.Appearance: SolidBoiling Point : 1386.05 °C at 760 mmHg (Predicted)Density : 1.43 g/cm3 (Predicted)Melting Point: 251-253 °CRTECS Number: AU1575000