Learn More
AdipoGen Actinomycin D,Chemical. CAS: 50-76-0
SDP
Catalog No. 50464830
Encompass_Preferred
Actinomycin D, AdipoGen Life Sciences. Chemical. CAS: 50-76-0. Formula: C62H86N12O16. MW: 1255.4. Isolated from Streptomyces parvulus. Antibiotic. Antitumor compound. Cytotoxic. Apoptosis inducer. RNA synthesis inhibitor. DNA intercalating agent. Mcl-1 expression inhibitor p53 pathway activator.
Catalog No. 50-464-830 Supplier AdipoGen Supplier No. BVT0089M005
Actinomycin D, AdipoGen Life Sciences. Chemical. CAS: 50-76-0. Formula: C62H86N12O16. MW: 1255.4. Isolated from Streptomyces parvulus. Antibiotic. Antitumor compound. Cytotoxic. Apoptosis inducer. RNA synthesis inhibitor. DNA intercalating agent. Mcl-1 expression inhibitor p53 pathway activator.