Learn More
Enzo Life Sciences Actinomycin D, (5mg), CAS Number: 50-76-0
SDP
Catalog No. NC9856244
Encompass_Preferred
Intercalating transcription inhibitor. An intercalating transcription inhibitor and antineoplastic antibiotic. Inhibits transcription by binding to DNA at dG residues. Induces apoptosis in many cell lines, but suppresses in PC12 cells induced by topoisomrease II inhibitor etopside. Used at 5-10µg/ml in cell culture for haltiong transcription. The product is not sterile. Formula: C62H86N12O16. MW: 1255.5. CAS Number: 50-76-0. Purity: ≥98% (TLC). Appearance: Red-orange crystalline solid. Solubility: Soluble in DMSO (>20mg/ml), 100% ethanol (1mg/ml) or methanol. Use/Stability: Sensitive to strong acids and bases. Store, as supplied, at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C
Catalog No. NC9856244 Supplier Enzo Life Sciences Supplier No. BMLGR3000005
Intercalating transcription inhibitor. An intercalating transcription inhibitor and antineoplastic antibiotic. Inhibits transcription by binding to DNA at dG residues. Induces apoptosis in many cell lines, but suppresses in PC12 cells induced by topoisomrease II inhibitor etopside. Used at 5-10µg/ml in cell culture for haltiong transcription. The product is not sterile. Formula: C62H86N12O16. MW: 1255.5. CAS Number: 50-76-0. Purity: ≥98% (TLC). Appearance: Red-orange crystalline solid. Solubility: Soluble in DMSO (>20mg/ml), 100% ethanol (1mg/ml) or methanol. Use/Stability: Sensitive to strong acids and bases. Store, as supplied, at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C