Learn More
Cayman Chemical ActInomycIn D 5mg
SDP
Catalog No. 502953640
Encompass_Preferred
A cyclic polypeptide-containing anticancer antibiotic that binds to DNA and blocks transcription by interfering with RNA synthesis; ~10 nM promotes cell death in a variety of tumors
Catalog No. 50-295-3640 Supplier Cayman Chemical Supplier No. 114215 MG
A cyclic polypeptide-containing anticancer antibiotic that binds to DNA and blocks transcription by interfering with RNA synthesis; ~10 nM promotes cell death in a variety of tumors