Learn More
Cayman Chemical Actinomycin D, 50-76-0, 5 mg
SDP
Supplier:  Cayman Chemical 114215
Encompass_Preferred
Molecular Formula C62H86N12O16, Purity ≥95%, Formula Weight 1255.5, Synonyms: 2-amino-N,N'-bis(hexadecahydro-6,13-diisopropyl-2,5,9-trimethyl-1,4,7,11,14-pentaoxo-1H-pyrrolo[2,1-i][1,4,7,10,13]oxatetraazacyclohexadecin-10-yl)-4,6-dimethyl-3-oxo-3H-phenoxazine-1,9-dicarboxamide, Cosmegen, Dactinomycin, Meractinomycin, NCI C04682, NSC 3053, Oncostatin K; A cyclic polypeptide-containing anticancer antibiotic that binds to DNA and blocks transcription by interfering with RNA synthesis; ~10 nM promotes cell death in a variety of tumors.
Catalog No. NC1379795
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.