Learn More
Carbosynth LLC. Actinomycin D, 5mg, CAS# 50-76-0, MF= C62H86N12O16
SDP
Supplier:  Carbosynth LLC. AMD3030PI5MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Dactinomycin, Application: A Double Cyclic Peptide-Containing Compound; Used As a Transcription Inhibitor; Used to Induce Apoptosis and Kill Tumor Cells, Notes: Bulk Pricing Available, origin/species: Streptomyces parvulus, one letter code: , MW: 1255.43
Catalog No. 50-195-2396