Learn More
AdipoGen Actinomycin D
SDP
Catalog No. 502562111
Encompass_Preferred
Chemical. CAS 50-76-0. Formula C62H86N12O16. MW 1255.4. Isolated from Streptomyces parvulus. Antibiotic. Antitumor compound. Cytotoxic. Apoptosis inducer. RNA synthesis inhibitor. DNA intercalating agent. Mcl-1 expression inhibitor p53 pathway activator.
Catalog No. 50-256-2111 Supplier AdipoGen Supplier No. BVT0089M025
Chemical. CAS 50-76-0. Formula C62H86N12O16. MW 1255.4. Isolated from Streptomyces parvulus. Antibiotic. Antitumor compound. Cytotoxic. Apoptosis inducer. RNA synthesis inhibitor. DNA intercalating agent. Mcl-1 expression inhibitor p53 pathway activator.