Learn More
Selleck Chemical LLC Actinomycin D S8964-10MG
SDP
Catalog No. 5000024501
Encompass_Preferred
Actinomycin D (Dactinomycin Act D RASP-101) is a significant polypeptide antibiotic isolated from soil bacteria of the genus Streptomyces Actinomycin D (Dactinomycin) inhibits DNA repair and rests the cell cycle at G1 phase with IC50 of 0 42 M and 0 4 nM respectively Actinomycin D is an autophagy activator induces p53-independent cell death and prolongs survival in high-risk chronic lymphocytic leukemia This product is a hazardous chemical (acute toxicity/flammable/skin corrosive) Please use it while wearing a protective face mask gloves and clothing
Catalog No. 50-000-24501 Supplier Selleck Chemical LLC Supplier No. S896410MG
Actinomycin D (Dactinomycin Act D RASP-101) is a significant polypeptide antibiotic isolated from soil bacteria of the genus Streptomyces Actinomycin D (Dactinomycin) inhibits DNA repair and rests the cell cycle at G1 phase with IC50 of 0 42 M and 0 4 nM respectively Actinomycin D is an autophagy activator induces p53-independent cell death and prolongs survival in high-risk chronic lymphocytic leukemia This product is a hazardous chemical (acute toxicity/flammable/skin corrosive) Please use it while wearing a protective face mask gloves and clothing