Learn More
Alkali Scientific Actinomycin D [Actinomycin C1] [Dactinomycin], 10 Milligrams
SDP
Catalog No. 5000661533
Encompass_Preferred
Actinomycin D, also known as dactinomycin, is a potent antibiotic and transcription inhibitor widely used in molecular and cellular biology research. It binds to DNA at guanine cytosine rich regions, preventing RNA polymerase from progressing during transcription. This effectively blocks RNA synthesis and is commonly used to study gene expression, transcription regulation, and cellular responses to DNA damage. Actinomycin D is also used in cancer research due to its ability to inhibit rapidly dividing cells. The compound is widely applied in laboratory experiments requiring precise control of RNA synthesis and gene expression processes.
Catalog No. 50-006-61533 Supplier Alkali Scientific Supplier No. ACT002CP
Actinomycin D, also known as dactinomycin, is a potent antibiotic and transcription inhibitor widely used in molecular and cellular biology research. It binds to DNA at guanine cytosine rich regions, preventing RNA polymerase from progressing during transcription. This effectively blocks RNA synthesis and is commonly used to study gene expression, transcription regulation, and cellular responses to DNA damage. Actinomycin D is also used in cancer research due to its ability to inhibit rapidly dividing cells. The compound is widely applied in laboratory experiments requiring precise control of RNA synthesis and gene expression processes.