Learn More
Alkali Scientific Actinomycin D [Actinomycin C1] [Dactinomycin], 5 Milligrams
SDP
Catalog No. 5000661532
Encompass_Preferred
Actinomycin D is a potent transcription inhibitor widely used in molecular biology and cancer research. It binds to DNA at guanine cytosine rich regions, preventing RNA polymerase from progressing during transcription. This effectively blocks RNA synthesis and is commonly used to study gene expression, transcriptional regulation, and cellular responses to DNA damage. Actinomycin D is also used in apoptosis studies and cancer cell research due to its ability to interfere with rapidly dividing cells. Supplied in a 5 milligram quantity, it is suitable for highly controlled laboratory experiments requiring precise inhibition of nucleic acid synthesis.
Catalog No. 50-006-61532 Supplier Alkali Scientific Supplier No. ACT001CP
Actinomycin D is a potent transcription inhibitor widely used in molecular biology and cancer research. It binds to DNA at guanine cytosine rich regions, preventing RNA polymerase from progressing during transcription. This effectively blocks RNA synthesis and is commonly used to study gene expression, transcriptional regulation, and cellular responses to DNA damage. Actinomycin D is also used in apoptosis studies and cancer cell research due to its ability to interfere with rapidly dividing cells. Supplied in a 5 milligram quantity, it is suitable for highly controlled laboratory experiments requiring precise inhibition of nucleic acid synthesis.