Learn More
Crescent Chemical Co Inc ACTINOMYCIN D CRYST.
SDP
Catalog No. 50247148
Encompass_Preferred
Actinomycin D cryst. research grade Chromopeptide with antibiotic activity. Contains 2 cyclic peptides bound to the chromophoric phenoxazone ring. Binds specifically to the minor groove of the DNA double helix, and thus prevents it from being a template for RNA
Catalog No. 50-247-148 Supplier Crescent Chemical Co Inc Supplier No. SE10710.01
Actinomycin D cryst. research grade Chromopeptide with antibiotic activity. Contains 2 cyclic peptides bound to the chromophoric phenoxazone ring. Binds specifically to the minor groove of the DNA double helix, and thus prevents it from being a template for RNA