Learn More
Sigma Aldrich Fine Chemicals Biosciences Actinomycin D from Streptomyces sp., ~98% (HPLC) | 50-76-0 | MFCD00005033 | 10MG
Catalog No. NC1591241
Encompass
Actinomycin D from Streptomyces sp., ~98% (HPLC) | Purity: ~98% (HPLC) | Mol Wt: 1255.42 | 50-76-0 | MFCD00005033 | 10MG
Catalog No. NC1591241 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A141010MG
Actinomycin D from Streptomyces sp., ~98% (HPLC) | Purity: ~98% (HPLC) | Mol Wt: 1255.42 | 50-76-0 | MFCD00005033 | 10MG