Learn More
Actinomycin D, USP Grade, G-Biosciences
SCI_ED
Catalog No. S139724
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139724 25 mg
S139792 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139724 Supplier G-Biosciences Supplier No. RC877
Only null left
Add to Cart
Add to Cart
Actinomycin D derives from Streptomyces species that inhibits cell proliferation by acting as a cytotoxic inducer of apoptosis against tumor cells.
It binds to DNA and inhibits RNA polymerase activity. Actinomycin is also used in plant tissue culture to inhibit plant growth through tissue deformation where it also acts as an RNA-synthesis inhibitor.
Specifications
Form Powder
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 25 mg