Learn More
AdipoGen Actinomycin X2
SDP
Catalog No. 502561966
Encompass_Preferred
Chemical. CAS 18865-48-0. Formula C62H84N12O17. MW 1269.4. Isolated from Streptomyces sp. Aniso-Actinomycin 4-Oxopro instead of Pro in the beta-chain. Antitumor and antiviral antibiotic. Antituberculosis agent. Has higher cytotoxicity towards cultured human leukemia HL-60 cells than actinomycin D. Apoptosis inducer. Forms a complex with DNA and blocks its biological activity.
Catalog No. 50-256-1966 Supplier AdipoGen Supplier No. BVT0375M001
Chemical. CAS 18865-48-0. Formula C62H84N12O17. MW 1269.4. Isolated from Streptomyces sp. Aniso-Actinomycin 4-Oxopro instead of Pro in the beta-chain. Antitumor and antiviral antibiotic. Antituberculosis agent. Has higher cytotoxicity towards cultured human leukemia HL-60 cells than actinomycin D. Apoptosis inducer. Forms a complex with DNA and blocks its biological activity.