Learn More
Medchemexpress LLC Actinonin | 13434-13-4 | 98.4% | 385.50 | 10 MG
SDP
Catalog No. 5000437903
Encompass_Preferred
Actinonin is a naturally occurring antibacterial agent produced by Actinomyces. It inhibits aminopeptidase M, aminopeptidase N, and leucine aminopeptidase. Actinonin is also a potent reversible peptide deformylase (PDF) inhibitor and inhibits several MMPs and hmeprin α. It induces apoptosis and exhibits antiproliferative and antitumor activities.
  • Inhibits aminopeptidase M, aminopeptidase N, and leucine aminopeptidase
  • Potent reversible peptide deformylase (PDF) inhibitor
  • Inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α
  • Induces apoptosis
  • Exhibits antiproliferative and antitumor activities
Catalog No. 50-004-37903 Supplier Medchemexpress LLC Supplier No. HY11395210MG
Actinonin is a naturally occurring antibacterial agent produced by Actinomyces. It inhibits aminopeptidase M, aminopeptidase N, and leucine aminopeptidase. Actinonin is also a potent reversible peptide deformylase (PDF) inhibitor and inhibits several MMPs and hmeprin α. It induces apoptosis and exhibits antiproliferative and antitumor activities.
  • Inhibits aminopeptidase M, aminopeptidase N, and leucine aminopeptidase
  • Potent reversible peptide deformylase (PDF) inhibitor
  • Inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α
  • Induces apoptosis
  • Exhibits antiproliferative and antitumor activities