Learn More
Medchemexpress LLC Actinonin | 13434-13-4 | 98.4% | 385.50 | 100 MG
SDP
Catalog No. 5000437125
Encompass_Preferred
Actinonin is a naturally occurring antibacterial agent produced by Actinomyces. It acts as an inhibitor of aminopeptidase M, aminopeptidase N, and leucine aminopeptidase. Actinonin is also a potent reversible peptide deformylase (PDF) inhibitor with a Ki of 0.28 nM. Additionally, it inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α with various Ki values. Actinonin induces apoptosis and demonstrates antiproliferative and antitumor activities.
  • Naturally occurring antibacterial agent produced by Actinomyces.
  • Inhibits aminopeptidase M, aminopeptidase N, and leucine aminopeptidase.
  • Potent reversible peptide deformylase (PDF) inhibitor with a Ki of 0.28 nM.
  • Inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α.
  • Acts as an apoptosis inducer.
  • Exhibits antiproliferative and antitumor activities.
Catalog No. 50-004-37125 Supplier Medchemexpress LLC Supplier No. HY113952100MG
Actinonin is a naturally occurring antibacterial agent produced by Actinomyces. It acts as an inhibitor of aminopeptidase M, aminopeptidase N, and leucine aminopeptidase. Actinonin is also a potent reversible peptide deformylase (PDF) inhibitor with a Ki of 0.28 nM. Additionally, it inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α with various Ki values. Actinonin induces apoptosis and demonstrates antiproliferative and antitumor activities.
  • Naturally occurring antibacterial agent produced by Actinomyces.
  • Inhibits aminopeptidase M, aminopeptidase N, and leucine aminopeptidase.
  • Potent reversible peptide deformylase (PDF) inhibitor with a Ki of 0.28 nM.
  • Inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α.
  • Acts as an apoptosis inducer.
  • Exhibits antiproliferative and antitumor activities.