Learn More
Medchemexpress LLC Actinonin | 13434-13-4 | 98.4% | 385.50 | 50 MG
SDP
Catalog No. 5000436065
Encompass_Preferred
Actinonin is a naturally occurring antibacterial agent produced by Actinomyces. It inhibits aminopeptidase M, aminopeptidase N, and leucine aminopeptidase, and is a potent reversible peptide deformylase (PDF) inhibitor (Ki: 0.28 nM). It also inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α, acting as an apoptosis inducer with antiproliferative and antitumor activities.
  • Naturally occurring antibacterial agent.
  • Inhibits aminopeptidases (M, N, leucine).
  • Potent reversible peptide deformylase (PDF) inhibitor.
  • Inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α.
  • Induces apoptosis.
  • Exhibits antiproliferative and antitumor activities.
  • Active against various gram-positive and gram-negative bacteria.
  • Solid appearance, white to off-white color.
  • Significant antitumor activity in prostate tumor xenograft model.
Catalog No. 50-004-36065 Supplier Medchemexpress LLC Supplier No. HY11395250MG
Actinonin is a naturally occurring antibacterial agent produced by Actinomyces. It inhibits aminopeptidase M, aminopeptidase N, and leucine aminopeptidase, and is a potent reversible peptide deformylase (PDF) inhibitor (Ki: 0.28 nM). It also inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α, acting as an apoptosis inducer with antiproliferative and antitumor activities.
  • Naturally occurring antibacterial agent.
  • Inhibits aminopeptidases (M, N, leucine).
  • Potent reversible peptide deformylase (PDF) inhibitor.
  • Inhibits MMP-1, MMP-3, MMP-8, MMP-9, and hmeprin α.
  • Induces apoptosis.
  • Exhibits antiproliferative and antitumor activities.
  • Active against various gram-positive and gram-negative bacteria.
  • Solid appearance, white to off-white color.
  • Significant antitumor activity in prostate tumor xenograft model.