Learn More
AdipoGen Actinonin, 13434-13-4, MFCD00080257, 5 mg
SDP
Supplier:  AdipoGen AGCN20161M005
Encompass_Preferred
MF: C19H35N3O5, MW: 385.5, >97.0%, Appearance: White solid. Antibiotic, Aminopeptidase M, N and leucine and alanine aminopeptidase inhibitor. Potent peptide deformylase (PDF) inhibitor. Anticancer compound. Antiproliferative. Shows antitumor activity in vitro and in vivo.
Catalog No. 50-595-743