Learn More
Superior Glove™ Action™ Aramid Fiber String Knit Cut-resistant Gloves, Grain-Leather Palm
Catalog No. 17489604
Change view
Click to view available options
Size:
Small
Catalog No. Size
17-489-604 Small
1 options
This item is not returnable. View return policy
Catalog No. 17-489-604 Supplier Superior Glove™ Supplier No. SKGLPS
This item is not returnable. View return policy
Cut-resistant aramid fiber gloves feature grain leather palm and thumb. Ideal for glass handling and metal stamping applications.
  • Leather palms wrap around sides of fingers, while thumb is all leather
  • Glove body made of knit Kevlar™ aramid fiber for cut and slash resistance, plus flame-resistance
  • Palms attached to knit shell using Kevlar™ aramid fiber thread for greater seam integrity
Specifications
Cut Resistance Rating ANSI A2
Material Aramid Fiber, Leather
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) sheet Metal, Metal stamping, Glass handling
Glove Cuff Style Knit