Learn More
Waters Corp Waters Corporation Activated Alumina, 0.2 kg, Mass Spectrometry Sorbent
Catalog No. 50792242 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This activated alumina is designed for use as a sorbent in foreline traps for mass spectrometry systems. It effectively absorbs oil mist and prevents the back-migration of rotary pump oil, ensuring optimal performance of your vacuum system.
  • Effectively absorbs oil mist.
  • Prevents back-migration of rotary pump oil.
  • Designed for mass spectrometry systems.
  • Suitable for use in foreline traps.
Catalog No. 50-792-242 Supplier Waters Corp Supplier No. 6070755
This activated alumina is designed for use as a sorbent in foreline traps for mass spectrometry systems. It effectively absorbs oil mist and prevents the back-migration of rotary pump oil, ensuring optimal performance of your vacuum system.
  • Effectively absorbs oil mist.
  • Prevents back-migration of rotary pump oil.
  • Designed for mass spectrometry systems.
  • Suitable for use in foreline traps.