Learn More
Perkin Elmer US LLC Activated Alumina, 0.45 kg
Catalog No. 502104115
Encompass_Preferred
For use in desiccants or an as absorbent. Can be used as a filter of fluoride, arsenic, and selenium in water. Highly porous material, excellent for absorbence.
Catalog No. 50-210-4115 Supplier Perkin Elmer US LLC Supplier No. 09923504
For use in desiccants or an as absorbent. Can be used as a filter of fluoride, arsenic, and selenium in water. Highly porous material, excellent for absorbence.