Learn More
Waters Corp Waters Activated Alumina, 0.45 kg
Catalog No. 50792158 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Activated Alumina is a highly adsorbent desiccant material, designed for efficient moisture removal in various laboratory and industrial applications. This product is particularly suitable for use in systems such as vacuum pump foreline traps, where its excellent drying capabilities help maintain optimal operating conditions.
  • Supplied in a 0.45 kg quantity.
  • Effectively removes moisture from air and gases.
  • Suitable for use in vacuum pump foreline traps.
Catalog No. 50-792-158 Supplier Waters Corp Supplier No. 6060107
Waters Activated Alumina is a highly adsorbent desiccant material, designed for efficient moisture removal in various laboratory and industrial applications. This product is particularly suitable for use in systems such as vacuum pump foreline traps, where its excellent drying capabilities help maintain optimal operating conditions.
  • Supplied in a 0.45 kg quantity.
  • Effectively removes moisture from air and gases.
  • Suitable for use in vacuum pump foreline traps.