Learn More
Strem, An Ascensus Company CAS# 7440-44-0. 2kg. Activated carbon. MFCD00133992
SDP
Catalog No. 5090108780
Encompass
CAS# 7440-44-0. 2kg. Activated carbon. MFCD00133992. Molecular Weight: 12.011. Molecular Formula: C. Color/form: 4 x 10 mesh black gran. Strem# 06-0050. http://www.strem.com/catalog/v/06-0050/
Catalog No. 50-901-08780 Supplier Strem, An Ascensus Company Supplier No. 0600502000G
CAS# 7440-44-0. 2kg. Activated carbon. MFCD00133992. Molecular Weight: 12.011. Molecular Formula: C. Color/form: 4 x 10 mesh black gran. Strem# 06-0050. http://www.strem.com/catalog/v/06-0050/