Learn More
LAB ALLEY Lab Alley Activated Carbon (Charcoal) Granular Food Grade Kosher
SDP
Catalog No. NC2402515
Encompass
Activated charcoal is a popular choice of adsorbent material for physical separation of impurities thanks to its high surface area and reactivity. It can effectively remove a wide range of pollutants including pesticides herbicides heavy metals phenols and chlorinated hydrocarbons. Granular activated charcoal is generally prepared via the physical or gas phase activation method. The food grade status of this product ensures that it may safely come into contact with food products.
Catalog No. NC2402515 Supplier LAB ALLEY Supplier No. ACCL1240G50LB
Activated charcoal is a popular choice of adsorbent material for physical separation of impurities thanks to its high surface area and reactivity. It can effectively remove a wide range of pollutants including pesticides herbicides heavy metals phenols and chlorinated hydrocarbons. Granular activated charcoal is generally prepared via the physical or gas phase activation method. The food grade status of this product ensures that it may safely come into contact with food products.