Learn More
Leica Biosystems Activated carbon filter - Replacement for Leica Peloris / Peloris II tissue processors
Supplier:  Leica Biosystems S26.0434
Encompass
Replacement activated carbon filter for Leica Biosystems Peloris and Peloris II tissue processors (Supplier part S26.0434). This filter adsorbs organic vapors and odors to protect equipment and maintain a clean processing environment.
  • Adsorbs organic vapors and odors.
  • Activated carbon media for high adsorption capacity.
  • Dimensions: 15 3/4 in x 7 3/8 in x 1 1/4 in.
  • Single filter element for easy replacement.
  • Helps protect internal components and reduce odors during processing.
Catalog No. NC1349503