Learn More
Leica Biosystems ACTIVATED CARBON FILTER FOR ST
Supplier: Leica Biosystems 14047432273
aCTIVaTED CaRBON FILTER - LEICa STaINING INSTRUMENTS (100 x 35 x 480mm) Equivalent to Leica Part Number: 14047432273 Compatible with: aEc Leica autoStainer XL (ST5010) aEc Leica MultiStainer (ST5020) aEc Leica Linear Stainer (ST4040) The manufacturer recommends that this filter is replaced at 3 monthly intervals (with normal usage