Learn More
Bioworld Granulated Activated Charcoal, 500 g
Catalog No. 50255193
Encompass_Preferred
High-purity carbon, especially useful for minimizing contamination in products. Supports or facilitates plant growth and/or shoot proliferation in two or more plant tissue cultures. This charcoal is plant tissue tested.Appearance: Grey solidDensity : 1.8-2.1g/cm3Oxidation_State: 4,3,2,1,0,-1,-2,-3,-4
Catalog No. 50-255-193 Supplier Bioworld Supplier No. 401000282
High-purity carbon, especially useful for minimizing contamination in products. Supports or facilitates plant growth and/or shoot proliferation in two or more plant tissue cultures. This charcoal is plant tissue tested.Appearance: Grey solidDensity : 1.8-2.1g/cm3Oxidation_State: 4,3,2,1,0,-1,-2,-3,-4